Potassium propionate 98%

Supplier: Ambeed
Warning

A785355-100G A785355-25G A785355-500G
76728-366EA 12 USD
76728-366 76728-368 76728-370
Potassium propionate 98%
Potassium propionate

Precaución: for Research Use Only

Formula: C₃H₅KO₂
MDL Number: MFCD00045936
CAS Number: 327-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR