N-(Benzyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: N-Benzylbenzenesulphonamide

A785045-100MG A785045-10G A785045-1G A785045-250MG A785045-5G
76728-356EA 29 USD
76728-356 76728-358 76728-360 76728-362 76728-364
N-(Benzyl)benzenesulfonamide 95%
N-(Benzyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂S
MW: 247.31 g/mol
MDL Number: MFCD00092709
CAS Number: 837-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR