1H-Indazol-1-amine 98%

Supplier: Ambeed
Warning

A783826-1G A783826-250MG
76728-348EA 185.66 USD
76728-348 76728-350
1H-Indazol-1-amine 98%
1H-Indazol-1-amine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD09910195
CAS Number: 33334-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR