N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 98%

Supplier: Ambeed
Warning

Synonyms: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt

A780045-100G A780045-25G A780045-500G A780045-5G A780045-1000G
76728-328EA 86 USD
76728-328 76728-330 76728-332 76728-334 77670-094
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 98%
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)

Precaución: for Research Use Only

Formula: C₁₉H₄₁NO₃S
MW: 363.5988 g/mol
MDL Number: MFCD00036910
CAS Number: 14933-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR