Potassium-5-methyl-2-thiophenetrifluoroborate 98%

Supplier: Ambeed
Warning

A779330-1G A779330-250MG A779330-5G
76728-318EA 39 USD
76728-318 76728-320 76728-322
Potassium-5-methyl-2-thiophenetrifluoroborate 98%
Potassium 5-methyl-2-thiophenetrifluoroborate

Precaución: for Research Use Only

Formula: C₅H₅BF₃KS
MDL Number: MFCD07784364
CAS Number: 871231-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR