2-Iodobenzophenone 97%

Supplier: Ambeed
Warning

A776198-1G A776198-25G A776198-5G A776198-100G
76728-294EA 17 USD
76728-294 76728-296 76728-298 77873-232
2-Iodobenzophenone 97%
2-Iodobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉IO
MDL Number: MFCD00270108
CAS Number: 25187-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR