Procarbazine hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: N-Isopropyl-4-[(2-methylhydrazino)methyl]benzamide, N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride, Procarbazine HCl

A775952-5G A775952-100MG A775952-1G A775952-250MG
76728-290EA 102 USD
76728-290 77602-988 77602-990 77880-554
Procarbazine hydrochloride 98%
Procarbazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O·HCl
MW: 257.76 g/mol
MDL Number: MFCD00072082
CAS Number: 366-70-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR