5-Methyl-DL-tryptophan 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH

A767222-100MG A767222-1G A767222-250MG A767222-5G
76728-230EA 19 USD
76728-230 76728-232 76728-234 76728-236
5-Methyl-DL-tryptophan 98%
5-Methyl-DL-tryptophan

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.26 g/mol
MDL Number: MFCD00005652
CAS Number: 951-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR