Sodium sulfanilate dihydrate sodium salt 97%

Supplier: Ambeed
Warning

A765574-1000G A765574-100G A765574-25G A765574-500G
76728-218EA 317.01 USD
76728-218 76728-220 76728-222 76728-224
Sodium sulfanilate dihydrate sodium salt 97%
Sodium sulfanilate Dihydrate

Precaución: for Research Use Only

Formula: C₆H₁₀NNaO₅S
MDL Number: MFCD00064394
CAS Number: 6106-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR