Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Di-(beta-carboethoxyethyl)methylamine

A760996-1G A760996-250MG
76728-208EA 11.67 USD
76728-208 76728-210
Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate 98%
Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
MDL Number: MFCD00026916
CAS Number: 6315-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR