4-Benzoylmorpholine 98%

Supplier: Ambeed
Warning

Synonyms: Morpholino(phenyl)methanone

A760954-100G A760954-1G A760954-25G A760954-5G
76728-196EA 158 USD
76728-196 76728-198 76728-200 76728-202
4-Benzoylmorpholine 98%
4-Benzoylmorpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.226 g/mol
MDL Number: MFCD00188446
CAS Number: 1468-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR