2-(Aminoethyl)-1-N-Boc-piperidine 95%

Supplier: Ambeed
Danger

A758320-100MG A758320-250MG
76728-180EA 28 USD
76728-180 77880-536
2-(Aminoethyl)-1-N-Boc-piperidine 95%
2-(Aminoethyl)-1-N-Boc-piperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD01631187
CAS Number: 239482-98-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR