Prima-1 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Bis(hydroxymethyl)-3-quinuclidinone, 2,2-Bis(hydroxymethyl)quinuclidin-3-one

A755385-10MG A755385-1MG A755385-25MG A755385-50MG A755385-5MG
76728-158EA 25.47 USD
76728-158 76728-160 76728-162 76728-164 76728-166
Prima-1 98%
Prima-1

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.223 g/mol
MDL Number: MFCD04974196
CAS Number: 5608-24-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR