p-Phenetidine hydrochloride 98%

Supplier: Ambeed
Warning

A754485-100G A754485-25G
76728-152EA 76 USD
76728-152 76728-154
p-Phenetidine hydrochloride 98%
p-Phenetidine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD00035503
CAS Number: 637-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR