Sodium pyridine-3-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Pyridine-3-sulphonic acid Sodium Salt

A752874-100G A752874-25G A752874-5G
76728-146EA 178 USD
76728-146 76728-148 76728-150
Sodium pyridine-3-sulfonate 98%
Sodium pyridine-3-sulfonate

Precaución: for Research Use Only

Formula: C₅H₅NO₃S
MDL Number: MFCD00065041
CAS Number: 15521-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR