Methyl-3,3,3-trifluoropropanate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 3,3,3-Trifluoropropionate, 3,3,3-Trifluoropropionic acid methyll ester

A752772-1G A752772-5G A752772-25G
76728-142EA 14.86 USD
76728-142 76728-144 77602-934
Methyl-3,3,3-trifluoropropanate 97%
Methyl-3,3,3-trifluoropropanate

Precaución: for Research Use Only

Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
MDL Number: MFCD02262206
CAS Number: 18830-44-9
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR