Disodium oxalate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

A751696-100G A751696-500G
76728-128EA 5 USD
76728-128 76728-130
Disodium oxalate 99%
Disodium oxalate

Precaución: for Research Use Only

Formula: Na₂C₂O₄
MDL Number: MFCD00012465
CAS Number: 62-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR