3-((Ethyl(phenyl)amino)methyl)benzenesulfonic acid 98%
Supplier: Ambeed
|
Warning
|
A747424-1000G
A747424-100G
A747424-25G
A747424-500G
76728-116EA
149
USD
76728-116
76728-118
76728-120
76728-122
3-((Ethyl(phenyl)amino)methyl)benzenesulfonic acid 98%
3-((Ethyl(phenyl)amino)methyl)benzenesulfonic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₇NO₃S |
MDL Number:
MFCD00035761 CAS Number: 101-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...