N-Carbobenzoxy-1,3-diaminopropane hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride

A746681-1G A746681-25G A746681-5G
76728-106EA 44.56 USD
76728-106 76728-108 76728-110
N-Carbobenzoxy-1,3-diaminopropane hydrochloride 95%
N-Carbobenzoxy-1,3-diaminopropane hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MW: 244.72 g/mol
MDL Number: MFCD00270153
CAS Number: 17400-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR