(R)-4-(Pyrrolidin-3-yl)morpholine 97+%

Supplier: Ambeed
Danger

A744420-100MG A744420-1G A744420-250MG
76728-100EA 70.02 USD
76728-100 76728-102 76728-104
(R)-4-(Pyrrolidin-3-yl)morpholine 97+%
(R)-4-(Pyrrolidin-3-yl)morpholine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O
MW: 156.22 g/mol
MDL Number: MFCD11519099
CAS Number: 511295-94-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR