2-Ethyl-2-phenylbutanoic acid 96%

Supplier: Ambeed
Warning

A743229-100MG A743229-1G A743229-250MG A743229-5G
76728-082EA 43.5 USD
76728-082 76728-084 76728-086 76728-088
2-Ethyl-2-phenylbutanoic acid 96%
2-Ethyl-2-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.25 g/mol
MDL Number: MFCD00075036
CAS Number: 5465-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR