Palladium(II) bromide 98%

Supplier: Ambeed
Warning

Synonyms: Palladium dibromide, Palladium bromide

A742182-1G A742182-250MG A742182-5G A742182-25G
76728-080EA 54 USD
76728-080 77602-898 77670-000 77880-517
Palladium(II) bromide 98%
Palladium(II) bromide

Precaución: for Research Use Only

Formula: Br₂Pd
MDL Number: MFCD00011170
CAS Number: 13444-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR