Phenoxyacetic anhydride 98%

Supplier: Ambeed
Warning

A741212-1G A741212-25G A741212-5G A741212-100G A741212-500G
76728-074EA 7 USD
76728-074 76728-076 76728-078 77552-274 77552-276
Phenoxyacetic anhydride 98%
Phenoxyacetic anhydride

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₅
MDL Number: MFCD00059681
CAS Number: 14316-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR