Silver(I) acetate 98%

Supplier: Ambeed
Danger

A740334-1G A740334-5G A740334-100G A740334-25G A740334-500G
76728-070EA 5 USD
76728-070 76728-072 77552-270 77552-272 77873-169
Silver(I) acetate 98%
Silver(I) acetate

Precaución: for Research Use Only

Formula: C₂H₃AgO₂
MDL Number: MFCD00012446
CAS Number: 563-63-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR