Methyl-4-fluoro-3-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-3-hydroxybenzoate

A739011-1G A739011-25G A739011-5G
76728-054EA 10.61 USD
76728-054 76728-056 76728-058
Methyl-4-fluoro-3-hydroxybenzoate 98%
Methyl 4-fluoro-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.14 g/mol
MDL Number: MFCD06797457
CAS Number: 214822-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR