Morpholino(piperidin-4-yl)methanone hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Piperidin-4-ylcarbonyl)morpholine hydrochloride

A738416-1G A738416-250MG A738416-5G
76728-042EA 106.09 USD
76728-042 76728-044 76728-046
Morpholino(piperidin-4-yl)methanone hydrochloride 98%
Morpholino(piperidin-4-yl)methanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₉ClN₂O₂
MW: 234.7231 g/mol
MDL Number: MFCD05864747
CAS Number: 94467-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR