N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), Ethylenediamine-N,N,N',N'-tetra-2-propanol

A737484-1000G A737484-100G A737484-500G
76728-028EA 49 USD
76728-028 76728-030 76728-032
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 98% (by GC)
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₄H₃₂N₂O₄
MW: 292.42 g/mol
MDL Number: MFCD00004534
CAS Number: 102-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR