N-Boc-3-(2-thienyl)-L-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-(2-thienyl)-Ala-OH, Boc-Thi-OH

A737431-1G A737431-250MG A737431-25G A737431-5G
76728-020EA 23 USD
76728-020 76728-022 76728-024 76728-026
N-Boc-3-(2-thienyl)-L-alanine 98%
N-Boc-3-(2-thienyl)-L-alanine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₄S
MW: 271.331 g/mol
MDL Number: MFCD00062051
CAS Number: 56675-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR