Piperitone 95% (by GC)

Supplier: Ambeed
Warning

A736891-1G A736891-25G A736891-5G
76728-014EA 10 USD
76728-014 76728-016 76728-018
Piperitone 95% (by GC)
Piperitone

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MDL Number: MFCD00045532
CAS Number: 89-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR