tert-Butyl (3-chloropropyl)carbamate 95%

Supplier: Ambeed
Warning

A734018-100MG A734018-1G A734018-250MG A734018-5G A734018-25G
76727-998EA 8 USD
76727-998 76728-000 76728-002 77569-384 77880-501
tert-Butyl (3-chloropropyl)carbamate 95%
tert-Butyl (3-chloropropyl)carbamate

Precaución: For research use only.

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD07781306
CAS Number: 116861-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR