Saquinavir mesylate 98%

Supplier: Ambeed
Warning

A732241-100MG A732241-10MG A732241-1MG A732241-25MG A732241-50MG
76727-982EA 94 USD
76727-982 76727-984 76727-986 76727-988 76727-990
Saquinavir mesylate 98%
Saquinavir mesylate

Precaución: for Research Use Only

Formula: C₃₈H₅₀N₆O₅·CH₄O₃S
MDL Number: MFCD00944907
CAS Number: 149845-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR