Bosutinib 99%

Supplier: Ambeed
Warning

Synonyms: SKI-606

A730879-100MG A730879-10MG A730879-1G A730879-1MG A730879-250MG A730879-50MG A730879-5G
76727-956EA 40.32 USD
76727-956 76727-958 76727-960 76727-962 76727-964 76727-966 76727-968
Bosutinib 99%
Bosutinib

Precaución: for Research Use Only

Formula: C₂₆H₂₉Cl₂N₅O₃
MW: 530.5 g/mol
MDL Number: MFCD07367846
CAS Number: 380843-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR