N-Carbobenzoxy-1,5-diaminopentane hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl (5-aminopentyl)carbamate hydrochloride, Benzyl N-(5-Aminopentyl)carbamate Hydrochloride, N-(5-Aminoamyl)carbamic Acid Benzyl Ester Hydrochloride, N-(5-Aminopentyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(5-Aminoamyl)carbamate Hydrochloride, N-Cbz-1,5-diaminopentane Hydrochloride

A728986-100MG A728986-250MG
76727-948EA 20.16 USD
76727-948 76727-950
N-Carbobenzoxy-1,5-diaminopentane hydrochloride 98%
N-Carbobenzoxy-1,5-diaminopentane hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 272.77 g/mol
MDL Number: MFCD00270152
CAS Number: 18807-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR