tert-Butyl 2-(2,5-dioxo-2H-pyrrol-1(5H)-yl)ethylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)carbamate, Mal-NH-Boc

A728155-100MG A728155-250MG A728155-1G A728155-5G
76727-940EA 17 USD
76727-940 76727-942 77602-880 77880-491
tert-Butyl 2-(2,5-dioxo-2H-pyrrol-1(5H)-yl)ethylcarbamate 97%
tert-Butyl 2-(2,5-dioxo-2H-pyrrol-1(5H)-yl)ethylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₄
MDL Number: MFCD03425523
CAS Number: 134272-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR