Pyridostigmine bromide 99+%

Supplier: Ambeed
Danger

Synonyms: 3-((Dimethylcarbamoyl)oxy)-1-methylpyridin-1-ium bromide, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide

A725388-1MG A725388-1G A725388-250MG
76727-920EA 6 USD
76727-920 77602-862 77669-954
Pyridostigmine bromide 99+%
Pyridostigmine bromide

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂O₂
MW: 261.12 g/mol
MDL Number: MFCD00079283
CAS Number: 101-26-8
UN: 2811
ADR: 6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR