Methyl-3-bromo-2-(bromomethyl)propionate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-(bromomethyl)propionic Acid Methyl Ester, Methyl Bis(bromomethyl)acetate, Methyl beta,beta'-Dibromoisobutyrate, Methyl 3-bromo-2-(bromomethyl)propanoate

A724892-100G A724892-1G A724892-25G A724892-5G
76727-912EA 515.6 USD
76727-912 76727-914 76727-916 76727-918
Methyl-3-bromo-2-(bromomethyl)propionate 97%
Methyl-3-bromo-2-(bromomethyl)propionate

Precaución: for Research Use Only

Formula: C₅H₈Br₂O₂
MW: 259.93 g/mol
MDL Number: MFCD00011534
CAS Number: 22262-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR