Pyrvinium Pamaote 99%

Supplier: Ambeed
Danger

Synonyms: 3-carboxy-1-[(3-carboxy-2-oxidonaphthalen-1-yl)methyl]naphthalen-2-olate, Pyrvinium pamoate, 2-(2-(2,5-Dimethyl-1-phenyl-1H-pyrrol-3-yl)vinyl)-6-(dimethylamino)-1-methylquinolin-1-ium bis-4,4'-methylenebis(3-hydroxy-2-naphthoate), 2-[(E)-2-(2,5-dimethyl-1-phenylpyrrol-3-yl)ethenyl]-N,N,1-trimethylquinolin-1-ium-6-amine

A722097-100MG A722097-10MG A722097-1MG A722097-50MG A722097-250MG
76727-900EA 37.14 USD
76727-900 76727-902 76727-904 76727-906 77669-948
Pyrvinium Pamaote 99%
Pyrvinium Pamaote

Precaución: for Research Use Only

Formula: C₇₅H₇₀N₆O₆
MW: 1151.42 g/mol
MDL Number: MFCD00010090
CAS Number: 3546-41-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR