4-(4-Nitrophenoxy)benzoic acid 98%

Supplier: Ambeed
Danger

A721902-100MG A721902-1G A721902-250MG A721902-5G
76727-892EA 16.98 USD
76727-892 76727-894 76727-896 76727-898
4-(4-Nitrophenoxy)benzoic acid 98%
4-(4-Nitrophenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₅
MW: 259.22 g/mol
MDL Number: MFCD00429494
CAS Number: 16309-45-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR