tert-Butyl-3-(aminomethyl)benzylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(aminomethyl)benzylcarbamate

A719533-100MG A719533-250MG A719533-25G A719533-5G
76727-878EA 15.92 USD
76727-878 76727-880 76727-882 76727-884
tert-Butyl-3-(aminomethyl)benzylcarbamate 97%
tert-Butyl-3-(aminomethyl)benzylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 236.32 g/mol
MDL Number: MFCD01317800
CAS Number: 108467-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR