N,N'-Dimethoxy-N,N'-dimethyloxamide 98%

Supplier: Ambeed
Danger

A718805-1G A718805-250MG A718805-25G A718805-5G
76727-866EA 61 USD
76727-866 76727-868 76727-870 77669-934
N,N'-Dimethoxy-N,N'-dimethyloxamide 98%
N,N'-Dimethoxy-N,N'-dimethyloxamide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄
MDL Number: MFCD02326951
CAS Number: 106675-70-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR