m-Toluenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A716381-1G A716381-25G A716381-5G
76727-832EA 11 USD
76727-832 76727-834 76727-836
m-Toluenesulfonyl chloride 98%
m-Toluenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₇ClO₂S
MDL Number: MFCD00051747
CAS Number: 1899-93-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR