Phenylphosphonic acid disodium salt hydrate 95%

Supplier: Ambeed
Warning

Synonyms: Benzenephosphonic Acid Disodium Salt

A715302-100G A715302-1G A715302-25G A715302-5G
76727-814EA 318 USD
76727-814 76727-816 76727-818 76727-820
Phenylphosphonic acid disodium salt hydrate 95%
Phenylphosphonic acid disodium salt hydrate

Precaución: for Research Use Only

Formula: C₆H₇O₃P
MDL Number: MFCD00058972
CAS Number: 25148-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR