Quinazoline 98%

Supplier: Ambeed
Warning

A714890-1G A714890-25G A714890-5G
76727-800EA 15 USD
76727-800 76727-802 76727-804
Quinazoline 98%
Quinazoline

Precaución: for Research Use Only

Formula: C₈H₆N₂
MDL Number: MFCD00006712
CAS Number: 253-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR