4-Pyrid-2-ylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Pyridyl)benzoic acid, 4-(Pyridin-2-yl)benzoic acid

A713884-1G A713884-250MG A713884-5G
76727-794EA 62.6 USD
76727-794 76727-796 76727-798
4-Pyrid-2-ylbenzoic acid 98%
4-Pyrid-2-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2100 g/mol
MDL Number: MFCD01830093
CAS Number: 4385-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR