N-Butylphthalimide 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Butylisoindoline-1,3-dione, N-(n-Butyl)phthalimide
A710193-100G
A710193-25G
A710193-5G
76727-774EA
51
USD
76727-774
76727-776
76727-778
N-Butylphthalimide 98+%
N-Butylphthalimide
Precaución: for Research Use Only
Formula:
C₁₂H₁₃NO₂ |
MDL Number:
MFCD00039695 CAS Number: 1515-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...