N-Chloroacetyl urethane 95%

Supplier: Ambeed
Danger

Synonyms: Ethyl (2-chloroacetyl)carbamate

A709868-1G A709868-250MG A709868-25G A709868-5G
76727-770EA 21 USD
76727-770 76727-772 77880-468 77880-469
N-Chloroacetyl urethane 95%
N-Chloroacetyl urethane

Precaución: for Research Use Only

Formula: C₅H₈ClNO₃
MW: 165.573 g/mol
MDL Number: MFCD00800542
CAS Number: 6092-47-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR