3-[4-(tert-Butyl)phenyl]acrylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butylcinnamic acid

A709519-1G A709519-5G A709519-250MG
76727-766EA 45 USD
76727-766 76727-768 77669-912
3-[4-(tert-Butyl)phenyl]acrylic acid 95%
3-[4-(tert-Butyl)phenyl]acrylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MDL Number: MFCD00040719
CAS Number: 1208-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR