Methyl-2-methyl-4-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-4-nitrobenzoate

A707734-100G A707734-10G A707734-1G A707734-25G A707734-500G A707734-5G
76727-744EA 140.04 USD
76727-744 76727-746 76727-748 76727-750 76727-752 76727-754
Methyl-2-methyl-4-nitrobenzoate 98%
Methyl 2-methyl-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.1700 g/mol
MDL Number: MFCD03618407
CAS Number: 62621-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR