1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 97%

Supplier: Ambeed
Warning

A706517-250MG A706517-1G
76727-736EA 67 USD
76727-736 77669-908
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 97%
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00040276
CAS Number: 4721-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR