2-((tert-Butoxycarbonyl)amino)-5-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A705823-100MG A705823-250MG
76727-730EA 71.09 USD
76727-730 76727-732
2-((tert-Butoxycarbonyl)amino)-5-(trifluoromethyl)benzoic acid 95%
2-((tert-Butoxycarbonyl)amino)-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄F₃NO₄
MW: 305.24 g/mol
MDL Number: MFCD09859761
CAS Number: 141940-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR